Assay Details
Target Gene Details
Entrez Gene ID: | 140609 |
Gene Name: | NIMA related kinase 7 |
Gene Aliases: |
- |
Location: |
Chr.1:198156978-198322420 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEK7 | NM_133494.2 | NP_598001.1 | ||
| XM_011509209.1 | XP_011507511.1 | |||
| XM_017000344.1 | XP_016855833.1 | |||
| XM_017000345.1 | XP_016855834.1 | |||
| XM_017000346.1 | XP_016855835.1 | |||
| XM_017000347.1 | XP_016855836.1 | |||
| XM_017000348.1 | XP_016855837.1 | |||
| AK313411.1 | ||||
| DB498485.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011794 | Chr.1:198055478 - 198190963 on Build GRCh38 | Gain |
|
| esv3588513 | Chr.1:198074849 - 198256653 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map