Assay Details
Target Gene Details
Entrez Gene ID: | 348378 |
Gene Name: | family with sequence similarity 159 member A |
Gene Aliases: |
PRO7171, WWLS2783 |
Location: |
Chr.1:52631438-52669120 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM159A | XM_011541377.2 | 1 | 355 | XP_011539679.1 |
| XM_011541378.2 | 1 | 356 | XP_011539680.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001535 | Chr.1:52620368 - 53346539 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80232329] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map