Assay Details
Target Gene Details
Entrez Gene ID: | 54810 |
Gene Name: | GIPC PDZ domain containing family member 2 |
Gene Aliases: |
SEMCAP-2, SEMCAP2 |
Location: |
Chr.1:78044962-78138449 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GIPC2 | NM_001304725.1 | NP_001291654.1 | ||
| NM_017655.5 | NP_060125.4 | |||
| AB073737.1 | BAB84711.1 | |||
| AK225941.1 | ||||
| AK307422.1 | ||||
| BC036075.1 | AAH36075.1 | |||
| BI561167.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78341252] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map