Assay Details
Target Gene Details
Entrez Gene ID: | 2590 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 2 |
Gene Aliases: |
GalNAc-T2 |
Location: |
Chr.1:230057789-230282130 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT2 | NM_001291866.1 | NP_001278795.1 | ||
| NM_004481.4 | NP_004472.1 | |||
| NR_120373.1 | ||||
| XM_017000963.1 | XP_016856452.1 | |||
| XM_017000965.1 | XP_016856454.1 | |||
| AK290048.1 | ||||
| AK296886.1 | ||||
| AK300453.1 | ||||
| AK304029.1 | ||||
| BC041120.1 | AAH41120.1 | |||
| BF727269.1 | ||||
| BG773173.1 | ||||
| X85019.1 | CAA59381.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832814 | Chr.1:230003613 - 230199636 on Build GRCh38 | Gain |
|
| esv3589083 | Chr.1:229939508 - 230186230 on Build GRCh38 | Gain |
|
| esv3589084 | Chr.1:229953231 - 230079036 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map