Assay Details
Target Gene Details
Entrez Gene ID: | 102724919 |
Gene Name: | uncharacterized LOC102724919 |
Gene Aliases: |
- |
Location: |
Chr.1:186435161-186470291 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724919 | NR_126002.1 | |||
| DB056828.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012164 | Chr.1:186354735 - 186596863 on Build GRCh38 | Gain |
|
| esv2762201 | Chr.1:186354747 - 186596875 on Build GRCh38 | Gain |
|
| esv3575361 | Chr.1:186266048 - 186604612 on Build GRCh38 | Gain |
|
| esv3890826 | Chr.1:185730915 - 186448352 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75036200] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map