Assay Details
Target Gene Details
Entrez Gene ID: | 1163 |
Gene Name: | CDC28 protein kinase regulatory subunit 1B |
Gene Aliases: |
CKS1, PNAS-16, PNAS-18, ckshs1 |
Location: |
Chr.1:154974642-154979249 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CKS1B | NM_001826.2 | NP_001817.1 | ||
| NR_024163.1 | ||||
| AF274943.1 | AAK07520.1 | |||
| AF279897.1 | AAK07553.1 | |||
| AK307066.1 | ||||
| AK311769.1 | ||||
| BC007751.2 | AAH07751.1 | |||
| BC015629.2 | AAH15629.1 | |||
| BC070319.1 | AAH70319.1 | |||
| BC070320.1 | AAH70320.1 | |||
| BQ883992.1 | ||||
| BT007196.1 | AAP35860.1 | |||
| CB161621.1 | ||||
| CN347951.1 | ||||
| X54941.1 | CAA38702.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map