Assay Details
Target Gene Details
Entrez Gene ID: | 100505832 |
Gene Name: | PROX1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:213819641-213986153 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PROX1-AS1 | NR_037850.2 | |||
| AK092251.1 |

Genomic Map