Assay Details
Target Gene Details
Entrez Gene ID: | 149420 |
Gene Name: | PDLIM1 interacting kinase 1 like |
Gene Aliases: |
CLIK1L, STK35L2 |
Location: |
Chr.1:26109406-26125548 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDIK1L | NM_001243532.1 | NP_001230461.1 | ||
| NM_001243533.1 | NP_001230462.1 | |||
| NM_152835.4 | NP_690048.1 | |||
| XM_005245743.4 | XP_005245800.1 | |||
| XM_006710381.3 | XP_006710444.1 | |||
| XM_011540792.2 | XP_011539094.1 | |||
| AF411102.1 | AAN03661.1 | |||
| AK312872.1 | ||||
| BC028713.1 | AAH28713.1 | |||
| CV024336.1 | ||||
| DA221039.1 | ||||
| DB227864.1 | ||||
| DB233669.1 |
More Information
Set Membership: |
|

Genomic Map