Assay Details
Target Gene Details
Entrez Gene ID: | 3176 |
Gene Name: | histamine N-methyltransferase |
Gene Aliases: |
HMT, HNMT-S1, HNMT-S2, MRT51 |
Location: |
Chr.2:137964068-138016364 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNMT | NM_001024075.1 | NP_001019246.1 | ||
| NM_006895.2 | NP_008826.1 | |||
| XM_011511064.2 | XP_011509366.1 | |||
| XM_017003948.1 | XP_016859437.1 | |||
| XM_017003949.1 | XP_016859438.1 | |||
| AF523356.1 | AAN33014.1 | |||
| AF523357.1 | AAN33015.1 | |||
| AF523358.1 | AAN33016.1 | |||
| AF523359.1 | AAN33017.1 | |||
| AF523360.1 | AAN33018.1 | |||
| AK301462.1 | ||||
| AK313804.1 | ||||
| BC020677.1 | AAH20677.1 | |||
| D16224.1 | BAA03752.1 | |||
| U08092.1 | AAA17423.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2671408 | Chr.2:137977575 - 137980222 on Build GRCh38 | Deletion |
|
| nsv583180 | Chr.2:137597152 - 138007481 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs138646677] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map