Assay Details
Target Gene Details
Entrez Gene ID: | 4249 |
Gene Name: | mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase |
Gene Aliases: |
GNT-V, GNT-VA |
Location: |
Chr.2:134119922-134454621 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGAT5 | XM_005263668.4 | XP_005263725.1 | ||
| XM_005263669.4 | XP_005263726.1 | |||
| XM_017004147.1 | XP_016859636.1 | |||
| XM_017004149.1 | XP_016859638.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7088n54 | Chr.2:134101185 - 134137859 on Build GRCh38 | Loss |
|
| nsv1008965 | Chr.2:133303486 - 134451957 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map