Assay Details
Target Gene Details
Entrez Gene ID: | 129446 |
Gene Name: | xin actin binding repeat containing 2 |
Gene Aliases: |
CMYA3 |
Location: |
Chr.2:166888487-167259753 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XIRP2 | NM_001079810.3 | NP_001073278.1 | ||
| NM_001199143.1 | NP_001186072.1 | |||
| NM_152381.5 | NP_689594.4 | |||
| AK097519.1 | ||||
| AK122889.1 | ||||
| AY820969.1 | AAV87913.1 | |||
| EF119718.1 | ||||
| EF119719.1 | ||||
| EF119720.1 | ||||
| EF119721.1 | ||||
| EF119722.1 |
Target Gene Details
Entrez Gene ID: | 100874011 |
Gene Name: | XIRP2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:167122809-167140955 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XIRP2-AS1 | NR_046665.1 | |||
| BU569857.1 | ||||
| DB050454.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3593278 | Chr.2:167120363 - 167169493 on Build GRCh38 | Loss |
|
| esv2672990 | Chr.2:167120347 - 167169476 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116674625] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map