Assay Details
Target Gene Details
Entrez Gene ID: | 115677 |
Gene Name: | nitric oxide synthase trafficking |
Gene Aliases: |
DaIP2 |
Location: |
Chr.2:168786539-168865339 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOSTRIN | NM_001171631.1 | NP_001165102.1 | ||
| AK093444.1 |
More Information
Set Membership: |
|

Genomic Map