Assay Details
Target Gene Details
Entrez Gene ID: | 10971 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein theta |
Gene Aliases: |
14-3-3, 1C5, HS1 |
Location: |
Chr.2:9583967-9631055 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YWHAQ | NM_006826.3 | NP_006817.1 | ||
| AF070556.1 | AAC28640.1 | |||
| AK297624.1 | ||||
| AK302208.1 | ||||
| AK309462.1 | ||||
| AK316158.1 | ||||
| BC050601.1 | AAH50601.1 | |||
| BC056867.1 | AAH56867.1 | |||
| BC093019.1 | AAH93019.1 | |||
| BT020014.1 | AAV38817.1 | |||
| X56468.1 | CAA39840.1 | |||
| X57347.1 | CAA40622.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv580915 | Chr.2:9168019 - 9704049 on Build GRCh38 | Gain |
|
| nsv963210 | Chr.2:9583405 - 9585499 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map