Assay Details
Target Gene Details
Entrez Gene ID: | 1123 |
Gene Name: | chimerin 1 |
Gene Aliases: |
ARHGAP2, CHN, DURS2, NC, RHOGAP2 |
Location: |
Chr.2:174799314-175005379 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHN1 | NM_001025201.3 | NP_001020372.2 | ||
| NM_001206602.1 | NP_001193531.1 | |||
| NM_001822.5 | NP_001813.1 | |||
| NR_038133.1 | ||||
| AB209788.1 | BAD93025.1 | |||
| AK055060.1 | ||||
| AK289941.1 | ||||
| AK300890.1 | ||||
| BC011393.1 | AAH11393.1 | |||
| CR456864.1 | CAG33145.1 | |||
| Z22641.1 | CAA80354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575313 | Chr.2:174591458 - 175085724 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map