Assay Details
Target Gene Details
Entrez Gene ID: | 10713 |
Gene Name: | ubiquitin specific peptidase 39 |
Gene Aliases: |
65K, CGI-21, HSPC332, SAD1, SNRNP65 |
Location: |
Chr.2:85602842-85649284 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP39 | NM_001256727.1 | 1 | 157 | NP_001243656.1 |
| NM_001256728.1 | 1 | 157 | NP_001243657.1 | |
| NR_046347.1 | ||||
| XM_006711922.1 | XP_006711985.1 | |||
| XM_011532488.1 | 1 | 138 | XP_011530790.1 | |
| AK056334.1 | ||||
| AK295257.1 | 1 | 155 | ||
| AK302236.1 | 1 | 134 | ||
| DA548921.1 | ||||
| DB444135.1 | 1 | 157 |
More Information
Set Membership: |
|

Genomic Map