Assay Details
Target Gene Details
Entrez Gene ID: | 57159 |
Gene Name: | tripartite motif containing 54 |
Gene Aliases: |
MURF, MURF-3, RNF30, muRF3 |
Location: |
Chr.2:27282429-27307439 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM54 | NM_032546.3 | NP_115935.3 | ||
| NM_187841.2 | NP_912730.2 | |||
| AJ291714.1 | CAC32841.1 | |||
| AK300699.1 | ||||
| BC141807.1 | ||||
| DA897004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833614 | Chr.2:27263559 - 27452213 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74564095] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map