Assay Details
Target Gene Details
Entrez Gene ID: | 51719 |
Gene Name: | calcium binding protein 39 |
Gene Aliases: |
CGI-66, MO25 |
Location: |
Chr.2:230712842-230821075 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAB39 | NM_001130849.1 | NP_001124321.1 | ||
| NM_001130850.1 | NP_001124322.1 | |||
| NM_016289.3 | NP_057373.1 | |||
| XM_011511350.2 | XP_011509652.1 | |||
| AK292382.1 | ||||
| AK315692.1 | ||||
| BC020570.1 | AAH20570.1 | |||
| BG773506.1 | ||||
| DB455871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584675 | Chr.2:230699911 - 230801231 on Build GRCh38 | Loss |
|
| nsv834565 | Chr.2:230697957 - 230920661 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76030306] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map