Assay Details
Target Gene Details
Entrez Gene ID: | 2736 |
Gene Name: | GLI family zinc finger 2 |
Gene Aliases: |
CJS, HPE9, PHS2, THP1, THP2 |
Location: |
Chr.2:120797291-120992653 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLI2 | NM_005270.4 | NP_005261.2 | ||
| XM_006712422.2 | XP_006712485.1 | |||
| XM_011510973.2 | XP_011509275.1 | |||
| XM_011510974.2 | XP_011509276.1 | |||
| AB007295.1 | BAA25665.1 | |||
| AB007296.1 | BAA25666.1 | |||
| AB007297.1 | BAA25667.1 | |||
| AB007298.1 | BAA25668.1 | |||
| AK300071.1 | ||||
| AY493737.1 | AAS72889.1 | |||
| AY493738.1 | AAS72890.1 | |||
| AY493739.1 | AAS72891.1 | |||
| BC111410.1 | ||||
| D14827.1 | BAA03568.1 | |||
| D14828.1 | BAA03569.1 | |||
| DQ004396.1 | AAY58315.1 | |||
| DQ004397.1 | AAY58316.1 | |||
| DQ004398.1 | AAY58317.1 | |||
| DQ086814.1 | AAY87165.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3592180 | Chr.2:120485127 - 121067968 on Build GRCh38 | Gain |
|
| nsv834349 | Chr.2:120765954 - 120967915 on Build GRCh38 | Loss |
|
| nsv834348 | Chr.2:120632010 - 120828123 on Build GRCh38 | Gain |
|
| nsv582798 | Chr.2:120552693 - 121587495 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115535313] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map