Assay Details
Target Gene Details
Entrez Gene ID: | 100506299 |
Gene Name: | ID2 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.2:8670984-8681863 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ID2-AS1 | NR_110153.1 | |||
| NR_110154.1 | ||||
| DA317685.1 | ||||
| HY143594.1 |

Genomic Map