Assay Details
Target Gene Details
Entrez Gene ID: | 25780 |
Gene Name: | RAS guanyl releasing protein 3 |
Gene Aliases: |
GRP3 |
Location: |
Chr.2:33436348-33564731 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASGRP3 | NM_170672.2 | NP_733772.1 | ||
| XM_011532746.2 | XP_011531048.1 | |||
| XM_017003759.1 | XP_016859248.1 | |||
| XM_017003762.1 | XP_016859251.1 | |||
| AK125551.1 | ||||
| AK292983.1 | ||||
| BC027849.1 | AAH27849.1 | |||
| DA439087.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762447 | Chr.2:33418657 - 33449417 on Build GRCh38 | Gain |
|
| esv3590286 | Chr.2:33415480 - 33446957 on Build GRCh38 | Loss |
|
| esv3891725 | Chr.2:33412332 - 33479056 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map