Assay Details
Target Gene Details
Entrez Gene ID: | 105373759 |
Gene Name: | uncharacterized LOC105373759 |
Gene Aliases: |
- |
Location: |
Chr.2:177211394-177214857 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105373759 | XM_017005483.1 | 2 | 2015 | XP_016860972.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834465 | Chr.2:177202566 - 177250385 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80351307] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map