Assay Details
Target Gene Details
Entrez Gene ID: | 51776 |
Gene Name: | sterile alpha motif and leucine zipper containing kinase AZK |
Gene Aliases: |
AZK, MLK7, MLT, MLTK, MRK, SFMMP, mlklak, pk |
Location: |
Chr.2:173075435-173268009 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZAK | XM_017004324.1 | XP_016859813.1 | ||
| AB041883.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3036 | Chr.2:173055503 - 173087887 on Build GRCh38 | Insertion |
|
| nsv834460 | Chr.2:173054736 - 173220560 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map