Assay Details
Target Gene Details
Entrez Gene ID: | 2591 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 3 |
Gene Aliases: |
GalNAc-T3, HFTC, HHS |
Location: |
Chr.2:165747803-165796352 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT3 | NM_004482.3 | NP_004473.2 | ||
| XM_005246449.1 | XP_005246506.1 | |||
| XM_011510929.1 | XP_011509231.1 | |||
| XM_017003770.1 | XP_016859259.1 | |||
| AK307391.1 | ||||
| AK313389.1 | ||||
| BC056246.1 | AAH56246.1 | |||
| BC113565.1 | ||||
| BC113567.1 | ||||
| X92689.1 | CAA63371.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583583 | Chr.2:165662006 - 165802076 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map