Assay Details
Target Gene Details
Entrez Gene ID: | 64172 |
Gene Name: | O-sialoglycoprotein endopeptidase-like 1 |
Gene Aliases: |
Qri7 |
Location: |
Chr.2:189746660-189763198 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OSGEPL1 | NM_022353.2 | NP_071748.2 | ||
| AK055441.1 | BAB70923.1 | |||
| AK314565.1 |
Target Gene Details
Entrez Gene ID: | 101409258 |
Gene Name: | OSGEPL1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:189762780-189765556 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OSGEPL1-AS1 | NR_102429.1 | |||
| BX474356.1 | ||||
| DB142079.1 |
More Information
Set Membership: |
|

Genomic Map