Assay Details
Target Gene Details
Entrez Gene ID: | 105373780 |
Gene Name: | proline-rich protein 18-like |
Gene Aliases: |
- |
Location: |
Chr.2:184593627-184598985 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105373780 | XM_011512285.1 | 3 | 748 | XP_011510587.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4114n100 | Chr.2:183277998 - 186056890 on Build GRCh38 | Gain |
|
| dgv232n21 | Chr.2:184301745 - 185428608 on Build GRCh38 | Loss |
|
| esv3893517 | Chr.2:182920870 - 185314087 on Build GRCh38 | Gain+Loss |
|

Genomic Map