Assay Details
Target Gene Details
Entrez Gene ID: | 150864 |
Gene Name: | family with sequence similarity 117 member B |
Gene Aliases: |
ALS2CR13 |
Location: |
Chr.2:202635178-202769757 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM117B | NM_173511.3 | NP_775782.2 | ||
| AB053315.1 | BAB69023.1 | |||
| AI523857.1 | ||||
| AK096090.1 | BAC04700.1 | |||
| CN397358.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7253n54 | Chr.2:202655549 - 202797056 on Build GRCh38 | Gain |
|
| nsv584179 | Chr.2:202663256 - 202768522 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map