Assay Details
Target Gene Details
Entrez Gene ID: | 100506108 |
Gene Name: | SIX3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:44940154-44941878 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIX3-AS1 | NR_103785.1 | 2 | 741 | |
| NR_103786.1 | 2 | 590 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526534 | Chr.2:44934159 - 44985100 on Build GRCh38 | Loss |
|
| nsv953779 | Chr.2:44932862 - 44955461 on Build GRCh38 | Deletion |
|
| nsv457207 | Chr.2:44931099 - 44967150 on Build GRCh38 | Loss |
|

Genomic Map