Assay Details
Target Gene Details
Entrez Gene ID: | 7850 |
Gene Name: | interleukin 1 receptor type 2 |
Gene Aliases: |
CD121b, CDw121b, IL-1R-2, IL-1RT-2, IL-1RT2, IL1R2c, IL1RB |
Location: |
Chr.2:101991805-102028544 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1R2 | NM_001261419.1 | NP_001248348.1 | ||
| NM_004633.3 | NP_004624.1 | |||
| NR_048564.1 | ||||
| XM_006712734.3 | XP_006712797.1 | |||
| XM_006712736.3 | XP_006712799.1 | |||
| XM_011511801.2 | 1 | 2178 | XP_011510103.1 | |
| XM_011511803.2 | XP_011510105.1 | |||
| XM_011511804.2 | XP_011510106.1 | |||
| XM_011511805.2 | XP_011510107.1 | |||
| XM_011511807.1 | 1 | 2178 | XP_011510109.1 | |
| XM_017004889.1 | XP_016860378.1 | |||
| AK130412.1 | ||||
| AK309161.1 | ||||
| BC027733.2 | AAH27733.1 | |||
| BC039031.1 | AAH39031.1 | |||
| DC380050.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs184563311,rs78485274] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map