Assay Details
Target Gene Details
Entrez Gene ID: | 100861402 |
Gene Name: | CERS6 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:168771951-168786429 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CERS6-AS1 | NR_045786.1 | |||
| NR_045787.1 | ||||
| DB072723.1 | ||||
| DB077980.1 |

Genomic Map