Assay Details
Target Gene Details
Entrez Gene ID: | 100507171 |
Gene Name: | BOLA3 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.2:74147981-74152389 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BOLA3-AS1 | NR_045634.1 | 2 | 4119 | |
| NR_045636.1 | 3 | 3993 |

Genomic Map