Assay Details
Target Gene Details
Entrez Gene ID: | 129446 |
Gene Name: | xin actin binding repeat containing 2 |
Gene Aliases: |
CMYA3 |
Location: |
Chr.2:166888487-167259753 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XIRP2 | NM_001079810.3 | NP_001073278.1 | ||
| NM_001199143.1 | NP_001186072.1 | |||
| NM_152381.5 | NP_689594.4 | |||
| AK097519.1 | ||||
| DB079087.1 | ||||
| EF119718.1 | ||||
| EF119719.1 | ||||
| EF119720.1 | ||||
| EF119721.1 | ||||
| EF119722.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3593257 | Chr.2:166648099 - 166928362 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map