Assay Details
Target Gene Details
Entrez Gene ID: | 100287879 |
Gene Name: | long intergenic non-protein coding RNA 994 |
Gene Aliases: |
- |
Location: |
Chr.3:64078361-64087363 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00994 | NR_033978.1 | |||
| AK125779.1 | BAC86286.1 |
Target Gene Details
Entrez Gene ID: | 100652759 |
Gene Name: | PRICKLE2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:64067964-64103131 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRICKLE2-AS1 | NR_045697.1 | |||
| AL832189.1 |

Genomic Map