Assay Details
Target Gene Details
Entrez Gene ID: | 4638 |
Gene Name: | myosin light chain kinase |
Gene Aliases: |
AAT7, KRP, MLCK, MLCK1, MLCK108, MLCK210, MSTP083, MYLK1, smMLCK |
Location: |
Chr.3:123612296-123884302 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYLK | NM_001321309.1 | NP_001308238.1 | ||
| NM_053025.3 | NP_444253.3 | |||
| NM_053026.3 | NP_444254.3 | |||
| NM_053027.3 | NP_444255.3 | |||
| NM_053028.3 | NP_444256.3 | |||
| XM_011512860.2 | XP_011511162.1 | |||
| XM_011512861.2 | XP_011511163.1 | |||
| XM_017006469.1 | XP_016861958.1 | |||
| XM_017006470.1 | XP_016861959.1 | |||
| XM_017006471.1 | XP_016861960.1 | |||
| AB037663.1 | BAB21504.1 | |||
| AF069601.2 | AAD15921.2 | |||
| AF069602.1 | AAD15922.1 | |||
| AF069603.1 | AAD15923.1 | |||
| AK300610.1 | ||||
| AY339601.1 | AAQ02673.1 | |||
| AY424269.1 | AAR29061.1 | |||
| AY424270.1 | AAR29062.1 | |||
| U48959.2 | AAC18423.2 | |||
| X85337.1 | CAA59685.1 |
Target Gene Details
Entrez Gene ID: | 100873940 |
Gene Name: | MYLK antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.3:123689644-123692407 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYLK-AS2 | NR_046629.1 | |||
| AI148949.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829710 | Chr.3:123641413 - 123825722 on Build GRCh38 | Gain |
|
| nsv591428 | Chr.3:123608828 - 124139902 on Build GRCh38 | Gain |
|
| nsv829708 | Chr.3:123519589 - 123690957 on Build GRCh38 | Gain |
|
| nsv829709 | Chr.3:123554281 - 123748120 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map