Assay Details
Target Gene Details
Entrez Gene ID: | 7029 |
Gene Name: | transcription factor Dp-2 |
Gene Aliases: |
DP2 |
Location: |
Chr.3:141944428-142149544 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv24180 | Chr.3:141965135 - 141966868 on Build GRCh38 | Loss |
|
| esv2669675 | Chr.3:141965120 - 141966999 on Build GRCh38 | Deletion |
|
| nsv1013352 | Chr.3:141822263 - 142151451 on Build GRCh38 | Gain |
|
| esv3597992 | Chr.3:141965106 - 141967010 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map