Assay Details
Target Gene Details
Entrez Gene ID: | 185 |
Gene Name: | angiotensin II receptor type 1 |
Gene Aliases: |
AG2S, AGTR1B, AT1, AT1AR, AT1B, AT1BR, AT1R, AT2R1, HAT1R |
Location: |
Chr.3:148697871-148743003 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGTR1 | NM_000685.4 | NP_000676.1 | ||
| NM_004835.4 | NP_004826.5 | |||
| NM_009585.3 | NP_033611.1 | |||
| NM_031850.3 | NP_114038.4 | |||
| AK291541.1 | ||||
| BC022447.1 | AAH22447.1 | |||
| BC068494.1 | AAH68494.1 | |||
| DA649921.1 | ||||
| M87290.1 | AAA35535.1 | |||
| M93394.1 | AAA58370.1 | |||
| S77410.1 | AAB34644.1 | |||
| X65699.1 | CAA46621.1 |
More Information
Set Membership: |
|

Genomic Map