Assay Details
Target Gene Details
Entrez Gene ID: | 90407 |
Gene Name: | transmembrane protein 41A |
Gene Aliases: |
2900010K02Rik |
Location: |
Chr.3:185482671-185499057 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM41A | NM_080652.3 | NP_542383.1 | ||
| XM_017007437.1 | XP_016862926.1 | |||
| AK172748.1 | BAD18735.1 | |||
| AK290878.1 | ||||
| AK313954.1 | ||||
| AY358547.1 | AAQ88911.1 | |||
| BC008043.2 | AAH08043.1 | |||
| BC019884.1 | AAH19884.1 | |||
| BP274550.1 | ||||
| BX647128.1 | ||||
| BX647928.1 | 2 | 520 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829818 | Chr.3:185411745 - 185567640 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map