Assay Details
Target Gene Details
Entrez Gene ID: | 100507032 |
Gene Name: | TMCC1 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.3:129893871-129908912 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMCC1-AS1 | NR_037893.1 | |||
| AK127796.1 |

Genomic Map