Assay Details
Target Gene Details
Entrez Gene ID: | 285441 |
Gene Name: | F11 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.4:186286098-186501058 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F11-AS1 | NR_033900.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1028146 | Chr.4:185582745 - 186417181 on Build GRCh38 | Gain |
|
| nsv461864 | Chr.4:186085652 - 186335850 on Build GRCh38 | Gain |
|
| nsv596401 | Chr.4:186151229 - 186445495 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72712626] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map