Assay Details
Target Gene Details
Entrez Gene ID: | 101929064 |
Gene Name: | uncharacterized LOC101929064 |
Gene Aliases: |
- |
Location: |
Chr.4:86119806-86219926 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929064 | NR_110879.1 |
Target Gene Details
Entrez Gene ID: | 5602 |
Gene Name: | mitogen-activated protein kinase 10 |
Gene Aliases: |
JNK3, JNK3A, PRKM10, SAPK1b, p493F12, p54bSAPK |
Location: |
Chr.4:86012296-86594117 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003585 | Chr.4:86036814 - 86329283 on Build GRCh38 | Loss |
|
| nsv1005338 | Chr.4:86102245 - 86187402 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map