Assay Details
Target Gene Details
Entrez Gene ID: | 132884 |
Gene Name: | EvC ciliary complex subunit 2 |
Gene Aliases: |
LBN, WAD |
Location: |
Chr.4:5531921-5709548 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EVC2 | XM_017007735.1 | 22 | 3821 | XP_016863224.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005173 | Chr.4:5503321 - 5536045 on Build GRCh38 | Gain |
|
| nsv1012934 | Chr.4:5534522 - 5568113 on Build GRCh38 | Gain |
|
| nsv522680 | Chr.4:5503406 - 5552407 on Build GRCh38 | Gain |
|
| nsv526713 | Chr.4:5527507 - 5552407 on Build GRCh38 | Gain |
|
| nsv593480 | Chr.4:4961200 - 5760434 on Build GRCh38 | Loss |
|
| nsv593482 | Chr.4:5184198 - 5593242 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map