Assay Details
Target Gene Details
Entrez Gene ID: | 126 |
Gene Name: | alcohol dehydrogenase 1C (class I), gamma polypeptide |
Gene Aliases: |
ADH3 |
Location: |
Chr.4:99336492-99353045 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADH1C | NM_000669.4 | NP_000660.1 | ||
| NR_133005.1 | ||||
| XM_011531588.2 | XP_011529890.1 | |||
| XM_011531589.1 | XP_011529891.1 | |||
| BC062476.1 | AAH62476.1 | |||
| BC066227.1 | AAH66227.1 | |||
| BC066228.1 | AAH66228.1 | |||
| BC067419.1 | AAH67419.1 | |||
| BC067420.1 | AAH67420.1 | |||
| BC067421.1 | AAH67421.1 | |||
| BC067422.1 | AAH67422.1 | |||
| BC074771.2 | AAH74771.1 | |||
| BC074786.2 | AAH74786.1 | |||
| CB114902.1 | ||||
| CD014078.1 | ||||
| M12272.1 | AAC41757.1 | |||
| X04299.1 | CAA27842.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5345n100 | Chr.4:99181544 - 99523288 on Build GRCh38 | Gain |
|
| nsv528518 | Chr.4:99336998 - 99353592 on Build GRCh38 | Loss |
|
| nsv1002809 | Chr.4:99312891 - 99364351 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map