Assay Details
Target Gene Details
Entrez Gene ID: | 51056 |
Gene Name: | leucine aminopeptidase 3 |
Gene Aliases: |
HEL-S-106, LAP, LAPEP, PEPS |
Location: |
Chr.4:17577304-17607967 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAP3 | NM_015907.2 | NP_056991.2 | ||
| AF061738.1 | AAD17527.1 | |||
| AK022055.1 | ||||
| AK130293.1 | ||||
| AK225511.1 | ||||
| AK298613.1 | ||||
| AK298790.1 | ||||
| BC006199.2 | AAH06199.3 | |||
| BC065564.1 | AAH65564.1 | |||
| CR457128.1 | CAG33409.1 | |||
| FJ224308.1 |
More Information
Set Membership: |
|

Genomic Map