Assay Details
Target Gene Details
Entrez Gene ID: | 1047 |
Gene Name: | calmegin |
Gene Aliases: |
- |
Location: |
Chr.4:140388453-140427661 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLGN | NM_001130675.1 | NP_001124147.1 | ||
| NM_004362.2 | NP_004353.1 | |||
| AK093096.1 | ||||
| AK290819.1 | ||||
| AK299243.1 | ||||
| AK302149.1 | ||||
| AL045602.1 | ||||
| BC028357.2 | AAH28357.1 | |||
| D86322.1 | BAA22590.1 | |||
| DC396076.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs116414613] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map