Assay Details
Target Gene Details
Entrez Gene ID: | 118 |
Gene Name: | adducin 1 |
Gene Aliases: |
ADDA |
Location: |
Chr.4:2843732-2930076 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3383963 | Chr.4:2877227 - 2879525 on Build GRCh38 | Duplication |
|
| dgv313n21 | Chr.4:2845072 - 2879529 on Build GRCh38 | Gain |
|
| esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion |
|
| nsv593435 | Chr.4:2769045 - 2879529 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map