Assay Details
Target Gene Details
Entrez Gene ID: | 255520 |
Gene Name: | ELMO domain containing 2 |
Gene Aliases: |
9830169G11Rik |
Location: |
Chr.4:140524146-140553770 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELMOD2 | NM_153702.3 | NP_714913.1 | ||
| XM_005262885.3 | XP_005262942.1 | |||
| XM_005262886.2 | XP_005262943.1 | |||
| XM_011531818.2 | 9 | 1244 | XP_011530120.1 | |
| XM_011531819.2 | XP_011530121.1 | |||
| AK312800.1 | ||||
| BC015168.1 | AAH15168.2 | |||
| BX648349.1 | ||||
| BX648843.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830088 | Chr.4:140436956 - 140603794 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map