Assay Details
Target Gene Details
Entrez Gene ID: | 5910 |
Gene Name: | Rap1 GTPase-GDP dissociation stimulator 1 |
Gene Aliases: |
GDS1, SmgGDS |
Location: |
Chr.4:98261376-98443861 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAP1GDS1 | NM_001100426.1 | NP_001093896.1 | ||
| NM_001100427.1 | NP_001093897.1 | |||
| NM_001100428.1 | NP_001093898.1 | |||
| NM_001100429.1 | NP_001093899.1 | |||
| NM_001100430.1 | NP_001093900.1 | |||
| NM_021159.4 | NP_066982.3 | |||
| AK055032.1 | ||||
| AK295156.1 | ||||
| BC001816.2 | AAH01816.1 | |||
| BC001851.2 | AAH01851.2 | |||
| BC098269.1 | AAH98269.1 | |||
| BC098334.1 | AAH98334.1 | |||
| BC099708.1 | AAH99708.1 | |||
| BC099845.1 | AAH99845.1 | |||
| DC367998.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528127 | Chr.4:98225285 - 98308688 on Build GRCh38 | Gain |
|
| esv2763801 | Chr.4:98173741 - 98492702 on Build GRCh38 | Gain |
|
| esv3575763 | Chr.4:98269773 - 98462320 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map