Assay Details
Target Gene Details
Entrez Gene ID: | 2028 |
Gene Name: | glutamyl aminopeptidase |
Gene Aliases: |
APA, CD249, gp160 |
Location: |
Chr.4:110476073-110563337 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENPEP | NM_001977.3 | NP_001968.3 | ||
| XM_017007877.1 | XP_016863366.1 | |||
| AK313412.1 | ||||
| AM888273.1 | ||||
| AM888274.1 | ||||
| AM888275.1 | ||||
| AM888276.1 | ||||
| AM888277.1 | ||||
| AM888278.1 | ||||
| AM888279.1 | ||||
| AM888280.1 | ||||
| AM888281.1 | ||||
| AM888282.1 | ||||
| AM888283.1 | ||||
| BC094770.1 | AAH94770.1 | |||
| BX641012.1 | ||||
| L12468.1 | AAA16876.1 | |||
| L14721.1 | AAA35522.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830036 | Chr.4:110485346 - 110674573 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map