Assay Details
Target Gene Details
Entrez Gene ID: | 84057 |
Gene Name: | meiotic nuclear divisions 1 |
Gene Aliases: |
GAJ |
Location: |
Chr.4:153344649-153415097 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MND1 | NM_001253861.1 | NP_001240790.1 | ||
| NM_032117.3 | NP_115493.1 | |||
| NR_045605.1 | ||||
| XM_005263275.2 | XP_005263332.1 | |||
| XM_017008688.1 | 1 | 401 | XP_016864177.1 | |
| AK312172.1 | ||||
| AY028916.1 | AAK26168.1 | |||
| BC032142.2 | AAH32142.1 | |||
| BF975093.1 | ||||
| BG613292.1 | ||||
| BG758047.1 | ||||
| BM820228.1 | ||||
| BM833450.1 |
More Information
Set Membership: |
|

Genomic Map