Assay Details
Target Gene Details
Entrez Gene ID: | 84142 |
Gene Name: | family with sequence similarity 175 member A |
Gene Aliases: |
ABRA1, CCDC98 |
Location: |
Chr.4:83459515-83485181 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM175A | NM_139076.2 | NP_620775.2 | ||
| XM_005263280.4 | XP_005263337.1 | |||
| XM_017008694.1 | XP_016864183.1 | |||
| AK022704.1 | BAB14189.1 | |||
| AK023676.1 | BAB14635.1 | |||
| AY358576.1 | AAQ88939.1 | |||
| BC039573.1 | AAH39573.1 | |||
| DA765424.1 | ||||
| EF531340.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893971 | Chr.4:83096792 - 83493129 on Build GRCh38 | Gain |
|
| nsv829994 | Chr.4:83445146 - 83637163 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map