Assay Details
Target Gene Details
Entrez Gene ID: | 4306 |
Gene Name: | nuclear receptor subfamily 3 group C member 2 |
Gene Aliases: |
MCR, MLR, MR, NR3C2VIT |
Location: |
Chr.4:148078764-148445287 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR3C2 | NM_000901.4 | NP_000892.2 | ||
| NM_001166104.1 | NP_001159576.1 | |||
| XM_011531975.1 | XP_011530277.1 | |||
| XM_011531976.2 | XP_011530278.1 | |||
| XM_011531977.2 | XP_011530279.1 | |||
| XM_017008217.1 | XP_016863706.1 | |||
| AB209056.1 | BAD92293.1 | |||
| AJ315514.1 | CAC67405.1 | |||
| AK304318.1 | ||||
| BC111758.1 | AAI11759.1 | |||
| M16801.1 | AAA59571.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3602613 | Chr.4:148081293 - 148109003 on Build GRCh38 | Loss |
|
| esv3602614 | Chr.4:148088251 - 148106596 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79358355] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map